missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyltrichlorosilane, 96%, Thermo Scientific™

$702.59 - $702.59

Chemical Identifiers

CAS 78560-44-8
Molecular Formula C10H4Cl3F17Si
Molecular Weight (g/mol) 581.55
MDL Number MFCD00042344
InChI Key VIFIHLXNOOCGLJ-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
PubChem CID 123577
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1658414
View Documents
Thermo Scientific Chemicals
L1658414
25 g
Each for $702.59
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltrichlorosilane is a halogenated compound used for biochemical research. It is applied to movable microparts of microelectromechanical systems , thereby reducing the surface energy and preventing the stickiness. Further, it is used to coat micro- and nano-features on stamps for a nanoimprint lithography. In addition, it is used in the preparation of (1H,1H,2H,2H-heptadecafluorodecyl)trimethoxysilane by reacting with methanol.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Moisture sensitive. Incompatible with oxidizing agents, strong acids and bases.
Specifications

Chemical Identifiers

78560-44-8
581.55
VIFIHLXNOOCGLJ-UHFFFAOYSA-N
123577
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
C10H4Cl3F17Si
MFCD00042344
1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane

Specifications

224°C
Liquid
C10H4Cl3F17Si
MFCD00042344
5889653
1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
VIFIHLXNOOCGLJ-UHFFFAOYSA-N
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
123577
96%
Moisture sensitive
1H,1H,2H,2H-Perfluorodecyltrichlorosilane
78560-44-8
25 g
CF3(CF2)7(CH2)2SiCl3
UN2987
>200°C (392°F)
Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents.
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
581.55
581.56
1.348
1.54
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'