missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%

Catalog No. AAB2407514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2407514 25 g
AAB2407506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2407514 Supplier Thermo Scientific Chemicals Supplier No. B2407514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2043-55-2 | C6H4F9I | 373.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2043-55-2
Molecular Formula C6H4F9I
Molecular Weight (g/mol) 373.988
MDL Number MFCD00039409
InChI Key CXHFIVFPHDGZIS-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
PubChem CID 74887
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
SMILES C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -25°C
Density 1.94
Boiling Point 138°C to 140°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.37
Quantity 25 g
Beilstein 1870862
Sensitivity Light sensitive
Formula Weight 373.99
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodohexane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-055-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only