missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-decanol, 97%, Thermo Scientific™

Catalog No. AAB2090209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2090209 10 g
AAB2090218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2090209 Supplier Thermo Scientific Chemicals Supplier No. B2090209

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a chemical applied to the formation of a barrier and self-healing coating on zinc metal materials. It induces cell death and formation of oxidative or reactive oxygen species in cerebellar granule cells. It is used to make surfactant and polymeric products with a range of commercial applications (may contain small amounts of residual unreacted material). The polymers are primarily used for coating textiles, paper, and carpet for oil, stain, and water repellency.

Solubility
Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 678-39-7
Molecular Formula C10H5F17O
Molecular Weight (g/mol) 464.122
MDL Number MFCD00039544
InChI Key JJUBFBTUBACDHW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro
PubChem CID 69619
ChEBI CHEBI:63919
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol
SMILES C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 45°C to 50°C
Boiling Point 112°C to 114°C (10 mmHg)
Quantity 10 g
Beilstein 2227487
Solubility Information Soluble in chloroform and methanol.
Formula Weight 464.12
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-decanol
EINECSNumber 211-648-0
TSCA Yes
Recommended Storage Ambient temperatures