missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Nonafluorohexyl Iodide 99.0+%, TCI America™
$67.38 - $167.49
Chemical Identifiers
| CAS | 2043-55-2 |
|---|---|
| Molecular Formula | C6H4F9I |
| Molecular Weight (g/mol) | 373.988 |
| MDL Number | MFCD00039409 |
| InChI Key | CXHFIVFPHDGZIS-UHFFFAOYSA-N |
| Synonym | 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane |
| PubChem CID | 74887 |
| IUPAC Name | 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane |
| SMILES | C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 2043-55-2 | |
| 373.988 | |
| CXHFIVFPHDGZIS-UHFFFAOYSA-N | |
| 74887 | |
| C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
| C6H4F9I | |
| MFCD00039409 | |
| 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane | |
| 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane |
Specifications
| 2043-55-2 | |
| Red | |
| MFCD00039409 | |
| 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane | |
| C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F | |
| 373.988 | |
| 373.99 | |
| Liquid |
| -25°C | |
| C6H4F9I | |
| 5 g | |
| CXHFIVFPHDGZIS-UHFFFAOYSA-N | |
| 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane | |
| 74887 | |
| ≥99.0% (GC) | |
| 1H,1H,2H,2H-Nonafluorohexyl Iodide |
Safety and Handling
TSCA : Yes