missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Nonafluorohexyl Iodide 99.0+%, TCI America™

Catalog No. P11555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11555G 5 g
P115525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11555G Supplier TCI America Supplier No. P11555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2043-55-2
Molecular Formula C6H4F9I
Molecular Weight (g/mol) 373.988
MDL Number MFCD00039409
InChI Key CXHFIVFPHDGZIS-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
PubChem CID 74887
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
SMILES C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -25°C
Color Red
Quantity 5 g
Formula Weight 373.99
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluorohexyl Iodide