missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Nonafluoro-1-hexanol 97.0+%, TCI America™

Catalog No. N06005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06005G 5 g
N060025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06005G Supplier TCI America Supplier No. N06005G
Only null left

Chemical Identifiers

CAS 2043-47-2
Molecular Formula C6H5F9O
Molecular Weight (g/mol) 264.09
MDL Number MFCD00039543
InChI Key JCMNMOBHVPONLD-UHFFFAOYSA-N
Synonym 2-(Perfluorobutyl)ethanol
PubChem CID 74883
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
SMILES OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 264.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluoro-1-hexanol
EINECSNumber (2)-2402
TSCA Yes