missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Nonafluoro-1-hexanol 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 76°C |
| Quantity | 25 g |
| Formula Weight | 264.09 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1H,1H,2H,2H-Nonafluoro-1-hexanol |
Safety and Handling
| EINECSNumber | (2)-2402 |
| TSCA | Yes |