missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized

Catalog No. AC348610050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348610050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC348610050 Supplier Thermo Scientific Chemicals Supplier No. 348610050

CAS: 27905-45-9 | C13H7F17O2 | 518.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27905-45-9
Molecular Formula C13H7F17O2
Molecular Weight (g/mol) 518.17
MDL Number MFCD00042306
InChI Key QUKRIOLKOHUUBM-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-heptadecafluorodecyl acrylate, 1h,1h,2h,2h-perfluorodecyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acrylate, unii-y9bfb4689z, 2-perfluorooctyl ethyl acrylate, 1,1,2,2-tetrahydroperfluorodecyl acrylate, perfluorooctyl ethyl acrylate, 2-propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester, 2-perfluorooctyl ethylacrylate, c8f17ch2ch2ococh=ch2
PubChem CID 119747
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
SMILES C=CC(=O)OCCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 100.0°C (4.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3360 to 1.3380
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, benzene, hexane
Stabilizer 90 to 110ppm MEHQ
Viscosity 12 mPa.s (25°C)
Formula Weight 518.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-722-7

RUO – Research Use Only