missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, tech. 90%, Thermo Scientific Chemicals
$197.71 - $197.71
Chemical Identifiers
| CAS | 183162-43-8 |
|---|---|
| Molecular Formula | C12H6F20O2 |
| Molecular Weight (g/mol) | 562.146 |
| MDL Number | MFCD00236625 |
| InChI Key | MBKZIVRAOJBYDI-UHFFFAOYSA-N |
| Synonym | 1h,1h,12h,12h-perfluoro-1,12-dodecanediol, 1h,1h,12h,12h-perfluorododecanediol, 1h,1h,12h,12h-perfluoro-1,12-dodecandiol, 1h,1h,12h,12h-perfluorododecane-1,12-diol, 1h,1h,12h,12h-icosafluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol |
| PubChem CID | 2776100 |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol |
| SMILES | C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1658806
|
Thermo Scientific Chemicals
L1658806 |
5 g |
Each for $197.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 183162-43-8 | |
| 562.146 | |
| MBKZIVRAOJBYDI-UHFFFAOYSA-N | |
| 2776100 | |
| C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| C12H6F20O2 | |
| MFCD00236625 | |
| 1h,1h,12h,12h-perfluoro-1,12-dodecanediol, 1h,1h,12h,12h-perfluorododecanediol, 1h,1h,12h,12h-perfluoro-1,12-dodecandiol, 1h,1h,12h,12h-perfluorododecane-1,12-diol, 1h,1h,12h,12h-icosafluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol |
Specifications
| 183162-43-8 | |
| 170°C (5 mmHg) | |
| MFCD00236625 | |
| 1h,1h,12h,12h-perfluoro-1,12-dodecanediol, 1h,1h,12h,12h-perfluorododecanediol, 1h,1h,12h,12h-perfluoro-1,12-dodecandiol, 1h,1h,12h,12h-perfluorododecane-1,12-diol, 1h,1h,12h,12h-icosafluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol | |
| C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 562.146 | |
| 562.15 | |
| Technical |
| 156°C to 160°C | |
| C12H6F20O2 | |
| 5 g | |
| MBKZIVRAOJBYDI-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol | |
| 2776100 | |
| 90% | |
| 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only