missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, tech. 90%, Thermo Scientific Chemicals

Catalog No. AAL1658806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1658806 Supplier Thermo Scientific Chemicals Supplier No. L1658806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 183162-43-8 | C12H6F20O2 | 562.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183162-43-8
Molecular Formula C12H6F20O2
Molecular Weight (g/mol) 562.146
MDL Number MFCD00236625
InChI Key MBKZIVRAOJBYDI-UHFFFAOYSA-N
Synonym 1h,1h,12h,12h-perfluoro-1,12-dodecanediol, 1h,1h,12h,12h-perfluorododecanediol, 1h,1h,12h,12h-perfluoro-1,12-dodecandiol, 1h,1h,12h,12h-perfluorododecane-1,12-diol, 1h,1h,12h,12h-icosafluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol
PubChem CID 2776100
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol
SMILES C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 156°C to 160°C
Boiling Point 170°C (5 mmHg)
Quantity 5 g
Formula Weight 562.15
Percent Purity 90%
Grade Technical
Chemical Name or Material 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only