missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol 98.0+%, TCI America™

Catalog No. I09941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09941G 1 g
I09945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09941G Supplier TCI America Supplier No. I09941G

Chemical Identifiers

CAS 183162-43-8
Molecular Formula C12H6F20O2
Molecular Weight (g/mol) 562.146
MDL Number MFCD00236625
InChI Key MBKZIVRAOJBYDI-UHFFFAOYSA-N
Synonym 1h,1h,12h,12h-perfluoro-1,12-dodecanediol, 1h,1h,12h,12h-perfluorododecanediol, 1h,1h,12h,12h-perfluoro-1,12-dodecandiol, 1h,1h,12h,12h-perfluorododecane-1,12-diol, 1h,1h,12h,12h-icosafluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol
PubChem CID 2776100
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol
SMILES C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 167°C
Color White
Boiling Point 170°C
Quantity 1 g
Formula Weight 562.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol