missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,10H,10H-Hexadecafluoro-1,10-decanediol 97.0+%, TCI America™

Catalog No. p-7130930
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12331G 1 g
H12335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12331G Supplier TCI America Supplier No. H12331G

Chemical Identifiers

CAS 754-96-1
Molecular Formula C10H6F16O2
Molecular Weight (g/mol) 462.13
MDL Number MFCD00192200
InChI Key NSKCTPBWPZPFHW-UHFFFAOYSA-N
Synonym 1h,1h,10h,10h-perfluorodecane-1,10-diol, 1h,1h,10h,10h-perfluoro-1,10-decanediol, 1h,1h,10h,10h-hexadecafluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decanediol, 1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, hexadecafluorooctamethylenebismethanol, 1,10-decanediol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecakis fluoranyl decane-1,10-diol
PubChem CID 2733404
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol
SMILES C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 138°C
Color White
Boiling Point 132°C
Quantity 1 g
Formula Weight 462.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H,10H,10H-Hexadecafluoro-1,10-decanediol