missing translation for 'onlineSavingsMsg'
Learn More

1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 95.0+%, TCI America™

Catalog No. O040025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O040025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O040025MG Supplier TCI America Supplier No. O040025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.158
MDL Number MFCD00792620
InChI Key LZMHWZHOZLVYDL-UHFFFAOYSA-N
Synonym 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
PubChem CID 1456
IUPAC Name [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
SMILES C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

Melting Point 162°C
Quantity 25 mg
Formula Weight 187.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one
RTECSNumber RO0913150
TSCA No
Recommended Storage Refrigerator