Learn More
SAI Infusion Technologies 1cc Zero Dead Volume Syringe, sterile 10/pk
Supplier:  SAI Infusion Technologies Z1CC
Encompass
This item has a minimum qty of 2 per supplier requirements.
Sterile 1cc syringe with zero dead volume. Conserve dosing solution and sample volume by eliminating any waste in the syringe hub. Sterile 10/pk. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
Catalog No. NC0186769
Only null left