missing translation for 'onlineSavingsMsg'
Learn More

(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol 98.0+%, TCI America™

Catalog No. p-7125379
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22755G 5 g
C227525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22755G Supplier TCI America Supplier No. C22755G
Only null left

Chemical Identifiers

CAS 50409-12-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00043062
InChI Key FSDSKERRNURGGO-UHFFFAOYSA-N
PubChem CID 230351
IUPAC Name cyclohexane-1,3,5-triol
SMILES C1C(CC(CC1O)O)O

Specifications

Melting Point 188°C
Color White
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol