Learn More
Anton Paar 180mm PTFE Filling Tube for DMA 35 Basic
Catalog No. NC0367539
Encompass
To fill sample into the measuring cell of the DMA 35 portable density meter, the filling tube is dipped directly into the sample container. It is recommended to exchange the filling tube on a regular basis.Suitable for use with Anton Paar density meter DMA 35 Basic Portable Density and Concentration Meter NC1488853
Catalog No. NC0367539 Supplier Anton Paar Supplier No. ANP 68527
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
To fill sample into the measuring cell of the DMA 35 portable density meter, the filling tube is dipped directly into the sample container. It is recommended to exchange the filling tube on a regular basis.Suitable for use with Anton Paar density meter DMA 35 Basic Portable Density and Concentration Meter NC1488853