missing translation for 'onlineSavingsMsg'
Learn More

18-Pentatriacontanone, tech. 85%

Catalog No. p-7051526
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4144514 25 g
AA4144522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4144514 Supplier Thermo Scientific Chemicals Supplier No. 04144514
Only null left

Antiblocking agent | CAS: 504-53-0 | C35H70O | 506.94 g/mol

18-Pentatriacontanone is used as ant blocking agent. It is also used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-53-0
Molecular Formula C35H70O
Molecular Weight (g/mol) 506.94
MDL Number MFCD00048491
InChI Key DMCJFWXGXUEHFD-UHFFFAOYSA-N
Synonym 18-pentatriacontanone, stearone, heptadecyl ketone, diheptadecyl ketone, di-n-heptadecyl ketone, unii-7in8ga6m68, stearon, stearyl ketone, di-stearyl ketone, acmc-1asrl
PubChem CID 10440
IUPAC Name pentatriacontan-18-one
SMILES CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 79°C to 80°C
Color White
Linear Formula (CH3(CH2)16)2CO
Quantity 25 g
Solubility Information Soluble in benzene
Formula Weight 506.94
Percent Purity 85%
Physical Form Powder
Chemical Name or Material 18-Pentatriacontanone
EINECSNumber 207-993-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only