missing translation for 'onlineSavingsMsg'
Learn More

18-Pentatriacontanone 95.0+%, TCI America™

Catalog No. p-7135056
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P129025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P129025G Supplier TCI America Supplier No. P129025G

Chemical Identifiers

CAS 504-53-0
Molecular Formula C35H70O
Molecular Weight (g/mol) 506.94
MDL Number MFCD00048491
InChI Key DMCJFWXGXUEHFD-UHFFFAOYSA-N
Synonym 18-pentatriacontanone, stearone, heptadecyl ketone, diheptadecyl ketone, di-n-heptadecyl ketone, unii-7in8ga6m68, stearon, stearyl ketone, di-stearyl ketone, acmc-1asrl
PubChem CID 10440
IUPAC Name pentatriacontan-18-one
SMILES CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 89°C
Color White
Boiling Point 345°C
Quantity 25 g
Formula Weight 506.94
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 18-Pentatriacontanone
EINECSNumber (2)-2475
TSCA Yes