Learn More
Chemglass Life Sciences 18 Gauge Needle, 6"" Long Metal Hubbed
Supplier: Chemglass Inc CG307540
18 Gauge Needle, 6" Long Metal Hubbed. Needle for CG-3070 and CG-3074 syringes. All needles are made of #304 full hard stainless steel and have a 22 beveled point for septum penetration along with general use. Removable needles come complete with the PTFE ferrule necessary for assembly.