missing translation for 'onlineSavingsMsg'
Learn More
Learn More
18-Crown 6-Ether 98.0+%, TCI America™
$57.75 - $510.16
Chemical Identifiers
| CAS | 17455-13-9 |
|---|---|
| Molecular Formula | C12H24O6 |
| Molecular Weight (g/mol) | 264.32 |
| MDL Number | MFCD00005113 |
| InChI Key | XEZNGIUYQVAUSS-UHFFFAOYSA-N |
| Synonym | 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane |
| PubChem CID | 28557 |
| ChEBI | CHEBI:32397 |
| IUPAC Name | 1,4,7,10,13,16-hexaoxacyclooctadecane |
| SMILES | C1COCCOCCOCCOCCOCCO1 |
Chemical Identifiers
| 17455-13-9 | |
| 264.32 | |
| XEZNGIUYQVAUSS-UHFFFAOYSA-N | |
| 28557 | |
| 1,4,7,10,13,16-hexaoxacyclooctadecane |
| C12H24O6 | |
| MFCD00005113 | |
| 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane | |
| CHEBI:32397 | |
| C1COCCOCCOCCOCCOCCO1 |
Specifications
| 17455-13-9 | |
| White-Yellow | |
| C12H24O6 | |
| 5 g | |
| XEZNGIUYQVAUSS-UHFFFAOYSA-N | |
| 1,4,7,10,13,16-hexaoxacyclooctadecane | |
| 28557 | |
| 264.32 | |
| Crystalline Powder |
| 40°C | |
| 190°C | |
| MFCD00005113 | |
| 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane | |
| C1COCCOCCOCCOCCOCCO1 | |
| 264.32 | |
| CHEBI:32397 | |
| ≥98.0% (GC) | |
| 18-Crown 6-Ether |
Safety and Handling
RTECSNumber : MP4500000
TSCA : Yes
Recommended Storage : Refrigerator