missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 16 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Description
Accommodate 16mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
Specifications
| Type | Ridge Tube Adapter |
| Tube Type | 16 mL Nalgene Oak Ridge Tube |
| Capacity | 16 mL |
| For Use With (Equipment) | Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors |
| Description | 16mL Nalgene Oak Ridge Tube Adapter |