Learn More
Electron Microscopy 16% Paraformaldehyde Aqueous Solution, EM Grade, Bottle 100 ML DFS Item
Supplier:  Electron Microscopy Sciences 15710S
Paraformaldehyde methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives.
Grade: EM Grade
Package: Bottle
Numéro de catalogue. 50980488
Il en reste null
Découvrez plus d'offres spéciales