missing translation for 'onlineSavingsMsg'
Learn More

16-Hexadecanolide 97.0+%, TCI America™

Catalog No. p-7130926
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
H12195G 5 g
H121925G 25 g
2 options

Catalog No. H12195G

Supplier: TCI America H12195G

Only null left

Chemical Identifiers

CAS 109-29-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00039668
InChI Key LOKPJYNMYCVCRM-UHFFFAOYSA-N
Synonym 16-hexadecanolide, hexadecanolide, cyclohexadecanolide, 16-hexadecanolactone, 1,16-hexadecanolide, dihydroambrettolide, juniperic acid lactone, 1,16-hexadecanolactone, hexadecanolactone, 16-hydroxyhexadecanoic acid lactone
PubChem CID 7984
IUPAC Name 1-oxacycloheptadecan-2-one
SMILES O=C1CCCCCCCCCCCCCCCO1

Specifications

Color Yellow
Boiling Point 188°C
Quantity 5 g
Formula Weight 254.41
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 16-Hexadecanolide
RTECSNumber RN9730000
TSCA Yes