missing translation for 'onlineSavingsMsg'
Learn More

16-Hexadecanolide, 97%

Catalog No. AAL0386409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0386409 10 g
AAL0386418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0386409 Supplier Thermo Scientific Chemicals Supplier No. L0386409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-29-5 | C16H30O2 | 254.41 g/mol

16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 109-29-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00039668
InChI Key LOKPJYNMYCVCRM-UHFFFAOYSA-N
Synonym 16-hexadecanolide, hexadecanolide, cyclohexadecanolide, 16-hexadecanolactone, 1,16-hexadecanolide, dihydroambrettolide, juniperic acid lactone, 1,16-hexadecanolactone, hexadecanolactone, 16-hydroxyhexadecanoic acid lactone
PubChem CID 7984
SMILES O=C1CCCCCCCCCCCCCCCO1

Specifications

Melting Point 33°C to 34°C
Flash Point >100°C (212°F)
Quantity 10 g
Beilstein 148631
Solubility Information Insoluble in water.
IUPAC Name 1-oxacycloheptadecan-2-one
Formula Weight 254.42
Percent Purity 97%
Chemical Name or Material 16-Hexadecanolide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 203-662-0
RTECSNumber RN9730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only