missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 15 mL Nunc Conical Tube or Filtration Devices Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor

Catalog No. 0101340
Catalog No. 0101340 Supplier Thermo Scientific™ Supplier No. 010 1340

Accommodate 15mL conical or filtration devices in Thermo Scientific™ Fiberlite™ conical tube rotors.

Accommodate 15mL Nunc conical tube or filtration devices in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.

Specifications

Type Filtration Devices Adapter
Tube Type Conical
Capacity 15 mL
For Use With (Equipment) Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description 15mL Nunc Conical Tube or Filtration Devices Adapter
Tube Size 15 mL