Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
GREENER_CHOICE
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
Specifications
Format Standard
Lid Without Lid
Array 32 x 48
No. of Wells 1536
Volume (Metric) Working 8 μL
View More Specs
Products 5
Catalog Number Mfr. No. Color Includes Price Quantity & Availability  
Catalog Number Mfr. No. Color Includes Price Quantity & Availability  
0720109
GREENER_CHOICE
View Documents Promotion Details
Corning™
3728BC
Black Generic barcodes N/A
Only null left
12456710
GREENER_CHOICE
View Documents Promotion Details
Corning™
4572
White, Clear Bottom Barcode N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12456706
GREENER_CHOICE
View Documents Promotion Details
Corning™
4567
Black, Clear Bottom Barcode N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
07200873
GREENER_CHOICE
View Documents Promotion Details
Corning™
C3895
Black, Clear Bottom Without barcode N/A
Only null left
0540001
GREENER_CHOICE
View Documents Promotion Details
Corning™
3728
Black Without barcode N/A
Only null left
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
Specifications
Specifications
Standard
32 x 48
8 μL
12.5 μL
50/Cs.
Without Lid
1536
Non-sterile
Flat Bottom (F-well)
10/Pk.