missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Nontreated Polystyrene Plates

Catalog No. 0720114
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
White
Volume (Metric) Working:
8 μL
10 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Working
0720114 Black, Clear Bottom 8 μL
07202510 Black 10 μL
07202511 White 8 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0720114 Supplier Corning™ Supplier No. 3891BC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Superior flatness minimizes variability within a plate and between plates

  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer™ FlexDrop™
  • Working volume of up to 10 μL
Generic Bar Code
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate No sequence number duplicates

Order Info

Packaged in bags of 10

Specifications

Barcode Includes Barcode
Format Standard
Lid Without Lid
Array 32 x 48
Color Black, Clear Bottom
For Use With (Equipment) Barcode scanning and software systems
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For fluorescent homogeneous assays, high throughput screening
Show More Show Less