missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 1536-well Cycloolefins Compound Storage Plate
GREENER_CHOICE

Catalog No. 07000909
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07000909 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000909 Supplier Greiner Bio-One Supplier No. 782855

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for acoustic liquid handling.

  • Free of detectable DNase, RNase, human DNA
  • Optimized tapered rounded square well design
  • Solid Flat bottom
  • Low dead volume
  • Deionized

Specifications

Array 32 x 48
Autoclavable Not Autoclavable
Color Clear
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working 1 to 10 μL
Sterility Non-sterile
Total Volume (Metric) Well 12 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Acoustic liquid handling
No. per Case 60
No. per Pack 15
Quantity 60/Cs.
RNase Free Yes
Temperature Range (Metric) -80°C to +100°C
Show More Show Less