Learn More
Chemglass Inc Reaction Vessel Lid, 5-Neck, 24/40 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange
Catalog No. CG194601
Encompass
Reaction Vessel Lid, 5-Neck, 24/40 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.
Catalog No. CG194601 Supplier Chemglass Inc Supplier No. CG194601
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reaction Vessel Lid, 5-Neck, 24/40 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.