Learn More
Pelican™ 1500 Medium Case
Medium cases provide mobility with room for protecting larger gear or multiple instruments and accessories.
Supplier:  Pelican™ 1500 005 150
 View more versions of this product
Catalog No. 19512732
Only null left
Description
Description
  • Watertight, dustproof case with easy open double throw latches
  • Made of ultra-high impact polypropylene
  • Multi layer lid organizer with numerous clean pockets
  • Lid organizer clear cover with Velcro™ fasteners
  • Padded dividers with adjustable walls for customizing compartments to any size
  • Stainless steel padlock protectors and three piece foam insert
  • Automatic pressure equalization valve and rubber over-molded handle
  • Lifetime guarantee of excellence
Certifications
IP67/MIL C-4150J/STANAG 4280/Def Stan 81-41/ATA 300
Specifications
Chemical Identifiers
6.12 in.
15.5 cm
18.5 in.
14 in.
28.4 cm
17.6 cm
18.5 x 14 in. (47 x 35.7 cm)
47 cm
35.7 cm
Specifications
EMS Case
Polypropylene
6.93 in.
17.6 cm
18.5 x 14 in. (47 x 35.7 cm)
Multi-layer lid organizer, padded dividers with adjustable walls
16.75 in.
42.5 cm
4.5 kg
11.1 in.
28.4 cm
Orange
Double throw latches, padlock protectors
6.12 in.
15.5 cm
Pressure equalization valve, dividers and lid organizer
18.5 in.
47 cm
10 lb.
14 in.
35.7 cm
Product Suggestions
Product Suggestions