Learn More
Mopec™ #15 Sterile Scalpel Blades
Catalog No. 22445418
Change view
Click to view available options
Blade Material:
Carbon Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Material
22445418 Carbon Steel
22445390 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22445418 Supplier Mopec™ Supplier No. AJ174
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-quality, sterile blades used for anatomic pathology or autopsy.
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Blade Material Carbon Steel
Product Type Scalpel
Size # 15
Disposable Single-use
Sterility Sterile
Quantity 100 Blades/Pk.
Graduated No