missing translation for 'onlineSavingsMsg'
Learn More

15-Hydroxypentadecanoic Acid 98.0+%, TCI America™

Catalog No. p-7130666
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06761G 1 g
H06765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06761G Supplier TCI America Supplier No. H06761G
Only null left

Chemical Identifiers

CAS 4617-33-8
Molecular Formula C15H30O3
Molecular Weight (g/mol) 258.402
MDL Number MFCD00039535
InChI Key BZUNJUAMQZRJIP-UHFFFAOYSA-N
PubChem CID 78360
ChEBI CHEBI:79169
IUPAC Name 15-hydroxypentadecanoic acid
SMILES C(CCCCCCCO)CCCCCCC(=O)O

Specifications

Melting Point 88°C
Color White
Quantity 1 g
Synonym 15-hydroxy-pentadecanoic acid, pentadecanoic acid, 15-hydroxy, acmc-1anlj, pentadecanoic acid,15-hydroxy, 15-hydroxypentadecanoic acid, 15-hydroxypentadecanoic acid capillary gc
Formula Weight 258.40
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 15-Hydroxypentadecanoic Acid
EINECSNumber (2)-1340
TSCA No