missing translation for 'onlineSavingsMsg'
Learn More

15-Hydroxypentadecanoic acid, 99+%

Catalog No. AAB2167003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2167003 1 g
AAB2167006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2167003 Supplier Thermo Scientific Chemicals Supplier No. B2167003

CAS: 4617-33-8 | C15H30O3 | 258.402 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4617-33-8
Molecular Formula C15H30O3
Molecular Weight (g/mol) 258.402
MDL Number MFCD00039535
InChI Key BZUNJUAMQZRJIP-UHFFFAOYSA-N
PubChem CID 78360
ChEBI CHEBI:79169
IUPAC Name 15-hydroxypentadecanoic acid
SMILES C(CCCCCCCO)CCCCCCC(=O)O

Specifications

Melting Point 84°C to 88°C
Linear Formula HO(CH2)14CO2H
Quantity 1 g
Beilstein 1240100
Synonym 15-hydroxy-pentadecanoic acid, pentadecanoic acid, 15-hydroxy, acmc-1anlj, pentadecanoic acid,15-hydroxy, 15-hydroxypentadecanoic acid, 15-hydroxypentadecanoic acid capillary gc
Formula Weight 258.4
Percent Purity ≥99%
Chemical Name or Material 15-Hydroxypentadecanoic acid
EINECSNumber 225-026-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only