missing translation for 'onlineSavingsMsg'
Learn More

15-Crown 5-Ether 97.0+%, TCI America™

Catalog No. p-7124649
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
C08595ML 5 mL
C085925ML 25 mL
2 options

Catalog No. C08595ML

Supplier: TCI America C08595ML

Only null left

Chemical Identifiers

CAS 33100-27-5
Molecular Formula C10H20O5
Molecular Weight (g/mol) 220.265
MDL Number MFCD00005110
InChI Key VFTFKUDGYRBSAL-UHFFFAOYSA-N
Synonym 15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
PubChem CID 36336
ChEBI CHEBI:32401
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadecane
SMILES C1COCCOCCOCCOCCO1

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 116°C
Quantity 5 mL
Formula Weight 220.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 15-Crown 5-Ether
RTECSNumber SB0200000
TSCA Yes