missing translation for 'onlineSavingsMsg'
Learn More

15-Crown-5, 98%

Catalog No. p-93783
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182860050 5 g Glass bottle
AC182860250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182860050 Supplier Thermo Scientific Chemicals Supplier No. 182860050
Only null left

CAS: 33100-27-5 | C10H20O5 | 220.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 33100-27-5
Molecular Formula C10H20O5
Molecular Weight (g/mol) 220.26
MDL Number MFCD00005110
InChI Key VFTFKUDGYRBSAL-UHFFFAOYSA-N
Synonym 15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
PubChem CID 36336
ChEBI CHEBI:32401
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadecane
SMILES C1COCCOCCOCCOCCO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 100.0°C to 135.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4640 to 1.4670
Quantity 5 g
Fieser 06,133; 07,76; 08,1280; 09,126; 12,147
Specific Gravity 1.1
Merck Index 14, 2602
Solubility Information Solubility in water: miscible
Formula Weight 220.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 15-Crown-5, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 251-379-6
RTECSNumber SB0200000
TSCA TSCA

RUO – Research Use Only