missing translation for 'onlineSavingsMsg'
Learn More

15-Azido-4,7,10,13-tetraoxapentadecanoic acid, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J64030MB

 View more versions of this product

Catalog No. AAJ64030MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

15-Azido-4,7,10,13-tetraoxapentadecanoic acid
C11H21N3O6
MFCD12406155
azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid
BODPHGOBXPGJKO-UHFFFAOYSA-N
22731907
1257063-35-6
N3CH2CH2(OCH2CH2)4CO2H
25mg
Soluble in water.
291.30
291.3

Chemical Identifiers

1257063-35-6
291.30
BODPHGOBXPGJKO-UHFFFAOYSA-N
22731907
C11H21N3O6
MFCD12406155
azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only