Learn More
World Precision Absorbent Paper Points, sterile, #15
Supplier:  World Precision WP504181
Encompass
Absorbent paper points are commonly used in laboratories, particularly in the field of dentistry and endodontics. These paper points are small, sterile, and highly absorbent. These sterile paper points are tapered to target specific areas and soak up small volumes of liquid. A variety of color-coded sizes are available, including extra fine, #15 fine, medium and coarse.
Catalog No. 50379946
Only null left