Learn More
Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper
Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.
$493.05 - $4197.60
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | COC |
| Closure Type | None |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Dimensions (L x W) | No. per Case | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | No. per Case | Price | Quantity | ||||
|
19540003
|
Keystone Cleanroom Products
T2ASHEET015X015 |
1.5 x 1.5 in. | 500 |
Case of 500 Each for $493.05
|
|
||||
|
19540004
|
Keystone Cleanroom Products
T2ASHEET0202 |
2 x 2 in. | 500 |
Case for $493.05
|
|
||||
|
19540005
|
Keystone Cleanroom Products
T2ASHEET0303 |
3 x 3 in. | 500 |
Case for $559.67
|
|
||||
|
19540006
|
Keystone Cleanroom Products
T2ASHEET0306 |
3 x 6 in. | 500 |
Case for $706.26
|
|
||||
|
19540007
|
Keystone Cleanroom Products
T2ASHEET0404 |
4 x 4 in. | 500 |
Case for $706.26
|
|
||||
|
19540008
|
Keystone Cleanroom Products
T2ASHEET0505 |
5 x 5 in. | 2000 |
Case for $3,637.91
|
|
||||
|
19540009
|
Keystone Cleanroom Products
T2ASHEET0606 |
6 x 6 in. | 1000 |
Case for $1,825.61
|
|
||||
|
19540010
|
Keystone Cleanroom Products
T2ASHEET0608 |
6 x 8 in. | 1000 |
Case for $2,105.46
|
|
||||
|
19540011
|
Keystone Cleanroom Products
T2ASHEET0609 |
6 x 9 in. | 1000 |
Case for $2,105.46
|
|
||||
|
19540012
|
Keystone Cleanroom Products
T2ASHEET0612 |
6 x 12 in. | 1000 |
Case for $2,385.30
|
|
||||
|
19540013
|
Keystone Cleanroom Products
T2ASHEET0624 |
6 x 24 in. | 500 |
Case for $2,385.30
|
|
||||
|
19540014
|
Keystone Cleanroom Products
T2ASHEET075075 |
7.5 x 7.5 in. | 1000 |
Case for $1,825.61
|
|
||||
|
19540015
|
Keystone Cleanroom Products
T2ASHEET0909 |
9 x 9 in. | 1000 |
Case for $2,105.46
|
|
||||
|
19540016
|
Keystone Cleanroom Products
T2ASHEET0911 |
9 x 11 in. | 500 |
Case for $1,119.36
|
|
||||
|
19540017
|
Keystone Cleanroom Products
T2ASHEET1010 |
10 x 10 in. | 500 |
Case for $1,119.36
|
|
||||
|
19540018
|
Keystone Cleanroom Products
T2ASHEET1212 |
12 x 12 in. | 1000 |
Case for $2,518.56
|
|
||||
|
19540020
|
Keystone Cleanroom Products
T2ASHEET1530 |
15 x 30 in. | 500 |
Case for $3,358.07
|
|
||||
|
19540021
|
Keystone Cleanroom Products
T2ASHEET1822 |
18 x 22 in. | 500 |
Case for $2,585.18
|
|
||||
|
19540022
|
Keystone Cleanroom Products
T2ASHEET2020 |
20 x 20 in. | 500 |
Case for $2,518.56
|
|
||||
|
19540023
|
Keystone Cleanroom Products
T2ASHEET2424 |
24 x 24 in. | 500 |
Case for $2,585.18
|
|
||||
|
19540024
|
Keystone Cleanroom Products
T2ASHEET3030 |
30 x 30 in. | 500 |
Case for $3,224.81
|
|
||||
|
19540025
|
Keystone Cleanroom Products
T2ASHEET3636 |
36 x 36 in. | 500 |
Case for $4,197.60
|
|
||||
|
19540027
|
Keystone Cleanroom Products
T2ASHEET4080 |
40 x 80 in. | 50 |
Case for $866.17
|
|
||||
|
19540028
|
Keystone Cleanroom Products
T2ASHEET4545 |
45 x 45 in. | 125 |
Case for $2,345.32
|
|
||||
|
19540029
|
Keystone Cleanroom Products
T2ASHEET4848 |
48 x 48 in. | 125 |
Case for $2,345.32
|
|
||||
|
19540030
|
Keystone Cleanroom Products
T2ASHEET5454 |
54 x 54 in. | 125 |
Case for $2,358.64
|
|
||||
|
19540031
|
Keystone Cleanroom Products
T2ASHEET5472 |
54 x 72 in. | 125 |
Case for $3,224.81
|
|
||||
Description
Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.
- Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
- Clean and consistently cut using an automated CAD system per ISO 8 Requirements
- Packaged in an ISO 6 Controlled Cleanroom
- Designed for Steam Sterilization
- Hydrophobic
- Resists tearing and punctures
Specifications
| Autoclavable | |
| None | |
| Single-use | |
| Pharmaceutical Production Equipment | |
| Non-sterile | |
| Yes | |
| <250°F | |
| Sterilization Wrap |
| COC | |
| White | |
| Steam Sterilization | |
| Tyvek | |
| 5.3 mm | |
| Steam, EtO, Gamma, VHP | |
| <125°C |