missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 13 mm HPLC and GC Vial Septa

Catalog No. 03377109
Change view
Click to view available options
Color:
Blue, Transparent
Red, White
Material:
PTFE, Silicone
Closure Size:
13-425
13-425 Screw Cap
Hardness:
30 Shore
45 Shore
50 Shore
55 Shore
57 °shore
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Closure Size Hardness
03377109 Red, White PTFE, Silicone 13-425 Screw Cap 45 Shore
03377107 Red, White PTFE, Silicone 13-425 Screw Cap 55 Shore
0334010G Red, White PTFE, Silicone 13-425 Screw Cap 50 Shore
03341947 Blue, Transparent PTFE, Silicone 13-425 Screw Cap 30 Shore
13622305 Red, White PTFE, Silicone 13-425 57 °shore
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03377109 Supplier Thermo Scientific™ Supplier No. C401545
Get 1 Extra Pack of Vials When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use Septa for 13-425 Screw Thread Caps to suit a variety of HPLC and GC applications

Choose Thermo Scientific 13 mm Septa for 13-425 Screw Thread Caps of PTFE, PTFE/Rubber or PTFE/Silicone to suit a variety of HPLC and GC applications.

  • PTFE disk septa are chemically inert
  • PFTE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles

Specifications

Description Septum for 13-425 Screw Caps
Color Red, White
Diameter (Metric) Outer 13 mm
Material PTFE, Silicone
Pre-slit No
Sterility Non-sterile
Disposable Single-use
Septum Silicone/PTFE
Packaging Type 1 pack of 100
Hardness 45 Shore
Quantity 100
Thickness (Metric) 1.5 mm
Closure Size 13-425 Screw Cap
For Use With (Equipment) 13-425 Screw Cap
Septa Size (Metric) 13 mm
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Product Type Septa
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.