missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 13 mm Autosampler Vial Screw Thread Caps

Catalog No. 03377108
Change view
Click to view available options
Color:
Black
Black, Blue, White
Black, Ivory, Red
Black, Red, White
Natural
Septum:
Ivory PTFE/Red Rubber
None
Polyethylene, Integral
PTFE/Silicone/PTFE
Silicone/PTFE
Material:
Phenolic
Polyethylene
Polypropylene
PTFE, Rubber, Rubber
PTFE, Silicone
PTFE, Silicone, Silicone
Product Type:
Open Top Screw Cap
Screw Cap
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Material Product Type
03377108 Black, Red, White PTFE/Silicone/PTFE PTFE, Silicone Open Top Screw Cap
0337525A Black None Polypropylene Open Top Screw Cap
03378315 Black None Phenolic Open Top Screw Cap
03378316 Black, Red, White Silicone/PTFE PTFE, Silicone Open Top Screw Cap
03377106 Black, Ivory, Red Ivory PTFE/Red Rubber PTFE, Rubber, Rubber Open Top Screw Cap
03376250 Black, Blue, White Silicone/PTFE PTFE, Silicone, Silicone Open Top Screw Cap
0337525C Natural Polyethylene, Integral Polyethylene Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03377108 Supplier Thermo Scientific™ Supplier No. C401540A
Get 1 Extra Pack of Vials When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.

Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.

  • Open-top
  • 13-425 GPI Thread

Specifications

Color (Seal) Red/White/Red
Description PP; Black; Red PTFE/White Silicone/Red PTFE septum
Color Black, Red, White
Material PTFE, Silicone
Sterility Non-sterile
Disposable Single-use
Septum PTFE/Silicone/PTFE
Quantity 100/Pk.
Thickness (Metric) 1.25 mm
Closure Size 13 mm
For Use With (Equipment) 13-425 Screw Thread Vials
Autoclavable Autoclavable
Certifications/Compliance Specification Certified
Product Type Open Top Screw Cap
Septum Color Red/White/Red
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.