missing translation for 'onlineSavingsMsg'
Learn More
Learn More
13-cis-Retinoic Acid 97.0+%, TCI America™
Supplier: TCI America R00881G
Specifications
| 13-cis-Retinoic Acid | |
| 175°C | |
| MFCD00079542 | |
| (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin | |
| CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C | |
| 300.442 | |
| 300.44 | |
| Crystalline Powder |
| 4759-48-2 | |
| C20H28O2 | |
| 1 g | |
| SHGAZHPCJJPHSC-RVDNBNLISA-N | |
| (2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid | |
| 131954766 | |
| ≥97.0% (HPLC,T) |
Chemical Identifiers
| 4759-48-2 | |
| 300.442 | |
| SHGAZHPCJJPHSC-RVDNBNLISA-N | |
| 131954766 | |
| CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C |
| C20H28O2 | |
| MFCD00079542 | |
| (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin | |
| (2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid |
Safety and Handling
RTECSNumber : VH6440000
TSCA : Yes
Recommended Storage : Freezer