missing translation for 'onlineSavingsMsg'
Learn More
Learn More
13-Azido-2,5,8,11-tetraoxatridecane 98.0+%, TCI America™
Supplier: TCI America A2728100MG
Specifications
| 13-Azido-2,5,8,11-tetraoxatridecane | |
| Yellow | |
| MFCD13184960 | |
| O-(2-Azidoethyl)-O′-methyltriethylene Glycol, mPEG4-Azide, mPEG4-N3, Methyl-PEG4-Azide | |
| COCCOCCOCCOCCN=[N+]=[N-] | |
| 233.268 | |
| 233.27 | |
| Liquid |
| 606130-90-9 | |
| C9H19N3O4 | |
| 100 mg | |
| FFOZZVDSANUDAE-UHFFFAOYSA-N | |
| 1-azido-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane | |
| 51340962 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 606130-90-9 | |
| 233.268 | |
| FFOZZVDSANUDAE-UHFFFAOYSA-N | |
| 1-azido-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane |
| C9H19N3O4 | |
| MFCD13184960 | |
| 51340962 | |
| COCCOCCOCCOCCN=[N+]=[N-] |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator