Malvern Instruments Inc 12mm Square Glass Cell with Square Aperture and Cap
Catalog No. NC0117958
12mm o.d. square glass cell with cap for size and electrophoresis measurements for the Zetasizer Nano & Zetasizer Advance series universal dip cell (ZEN1002)
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12mm o.d. square glass cell with cap for size and electrophoresis measurements for the Zetasizer Nano & Zetasizer Advance series universal dip cell (ZEN1002)