Learn More
Anton Paar 12 ml glass vials (100 pcs)
Supplier:  Anton Paar ANP 94197
Encompass
Pack of 100 glass vials used with a sample changer (Anton Paar Xsample series) to automatically fill up to 12 mL of sample into your Anton Paar measuring instrument.Recommended Cap Standard Cap for 12 ml Vials NC9145272 or High Temperarure Cap for 12 ml Vials NC9035965
Catalog No. NC1447533
Only null left