Learn More
Chemglass Inc 12000mL Glas-Col Heating Mantle, Series O, Hemispherical, 2 @ 650 Watts
Catalog No. CG1000012
Encompass
12000mL Glas-Col Heating Mantle, Series O, Hemispherical, 2 @ 650 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.
Catalog No. CG1000012 Supplier Chemglass Inc Supplier No. CG1000012
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12000mL Glas-Col Heating Mantle, Series O, Hemispherical, 2 @ 650 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.