Learn More
Mopec™ #12B Sterile Scalpel Blades
Catalog No. 22445405
Change view
Click to view available options
Blade Material:
Carbon Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Material
22445405 Stainless Steel
22445462 Carbon Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22445405 Supplier Mopec™ Supplier No. AJ206
Only null left
High-quality, sterile blades used for anatomic pathology or autopsy.
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Blade Material Stainless Steel
Product Type Scalpel
Size # 12B
Disposable Single-use
Sterility Sterile
Quantity 100 Blades/Pk.
Graduated No