Learn More
Integra Biosciences Ag Sterile, Filter, Low Retention, 125 ul Standard Tips, 5 XYZ Racks of 384 tips, 1920/CS
Catalog No. NC1341329
Encompass
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.
Catalog No. NC1341329 Supplier Integra Biosciences Ag Supplier No. ITG6565
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.