Learn More
Foxx Life Sciences EZBio Single Use Media Bottle Assembly, 125mL Sterilized PETG Bottles for Aseptic Sampling and Sterile Storage, 38-430 VersaCap with Vent and Dip Tube, 10/CS
Supplier: Foxx Life Sciences 4370003FLS
437-0003-FLS: EZBio, Single Use Bottle Assembly, 125mL, 38-430 VersaCap, PETG, Vented w/ DipTube, 10/cs. Foxx Life Sciences EZbio Single Use Sterile Media Bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components and Gamma Irradiated in the United States. Assemblies take advantage of Foxx patented VersaCap technology which means there is no machining necessary to produce the product. Perfect for single use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate. Foxx also offers the ability to provide and produce custom components, assemblies, and variations in a timely manner. For more information please contact us at sales@foxxlifesciences.com or by phone at 1-603-890-3699.