Ace Glass Clamp, two-piece, anodized, fits 137mm OD x 100mm ID flat flanges (500-1000mL reaction flasks).
Catalog No. 509557328
124mm two-piece anodized clamp for use in securing 500-1000mL reaction flasks. Black anodized, high-strength aluminum clamp has three wing nuts to secure flask to head. For use with 100mm I.D. flanges.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
124mm two-piece anodized clamp for use in securing 500-1000mL reaction flasks. Black anodized, high-strength aluminum clamp has three wing nuts to secure flask to head. For use with 100mm I.D. flanges.