missing translation for 'onlineSavingsMsg'
Learn More

Pelican™ 1200 Small Case

Catalog No. 17985414B
Encompass
Change view
Click to view available options
Color:
Black
Silver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17985414B Silver
17985414A Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985414B Supplier Pelican™ Supplier No. 1200 000 180

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lightweight cas is extremely strong, compact, and fully capable.

  • Watertight, crushproof, and dustproof
  • Easy open Double Throw latches
  • Open cell core with solid wall design makes the case strong yet light weight
  • O-ring seal
  • Automatic Pressure Equalization Valve balances interior pressure, keeps water out
  • Stainless steel hardware
  • Pick N Pluck™ with convoluted lid foam
  • Lifetime Guarantee of Excellence

Certifications

IP67/TANAG 4280/Def Stan 81-41

Specifications

Product Type Watertight Equipment Case
Color Silver
Material Polypropylene, Polyurethane Foam
Depth (English) 4.87 in.
Depth (English) Interior 4.12 in.
Depth (Metric) 12.4 cm
Depth (Metric) Interior 10.5 cm
Dimensions (L x W) 10.62 x 9.68 in. (27 x 24.6 cm)
Insert Type Pick N' Pluck
Length (English) 10.62 in.
Length (English) Interior 9.25 in.
Length (Metric) 27 cm
Length (Metric) Interior 23.5 cm
Model 1200
Weight (English) 2.45 lb.
Weight (Metric) 1.2 kg
Width (English) 9.68 in.
Width (English) Interior 7.12 in.
Width (Metric) 24.6 cm
Width (Metric) Interior 18.1 cm
Show More Show Less